Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

363189

Sigma-Aldrich

2-Bromobenzyl succinimidyl carbonate

97%

Synonym(s):

N-(2-Bromobenzyloxycarbonyloxy)succinimide, Z(2-Br)-OSu, Z(2-Br)-OSu, N-(2-Bromobenzyloxycarbonyloxy)succinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10BrNO5
CAS Number:
Molecular Weight:
328.12
Beilstein:
3623239
MDL number:

Assay

97%

SMILES string

Brc1ccccc1COC(=O)ON2C(=O)CCC2=O

InChI

1S/C12H10BrNO5/c13-9-4-2-1-3-8(9)7-18-12(17)19-14-10(15)5-6-11(14)16/h1-4H,5-7H2

InChI key

SZDNRTVADOCWKU-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service