Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

362433

Sigma-Aldrich

Diethyl (N-methoxy-N-methylcarbamoylmethyl)phosphonate

96%

Synonym(s):

N-Methoxy-N-methyl-2-(diethyl phosphono)acetamide, N-Methoxy-N-methyl-phosphonoacetamide diethyl ester, Diethyl N-methoxy-N-methylphosphonoacetamide, Diethylphosphonoacetic acid N-methyl-N-methoxyamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3ON(CH3)COCH2P(O)(OC2H5)2
CAS Number:
Molecular Weight:
239.21
Beilstein:
1875865
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.455 (lit.)

bp

90 °C/17 mmHg (lit.)

density

1.163 g/mL at 25 °C (lit.)

functional group

amine
phosphonate

SMILES string

CCOP(=O)(CC(=O)N(C)OC)OCC

InChI

1S/C8H18NO5P/c1-5-13-15(11,14-6-2)7-8(10)9(3)12-4/h5-7H2,1-4H3

InChI key

WYLRYBDGIILFIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of ring-expanded bryostatin analogs as antitumor agents
  • Asymmertic synthesis of pinnatoxins A and G via diastereoselective Ireland-Claisen rearrangement, ring closing metathesis, and cyclization
  • Rh2(II)-catalyzed nitro-group migration reactions
  • Stereoselective synthesis of cyclohexanones via phase transfer catalyzed double addition reactions
  • Stereoselective synthesis of (+)-polyrhacitide A via aldol reaction, Horner-Wardsworth-Emmons reaction, Evans acetal intramolecular oxa-Michael reaction and diastereoselective syn reduction reaction as the key steps
  • Preparation of substituted furans via olefin cross-metathesis
Wittig carbonylating reagent.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldrichimica Acta, 23, 43-43 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service