Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

361321

Sigma-Aldrich

1-(2-Methoxyphenyl)-2-thiourea

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4NHCSNH2
CAS Number:
Molecular Weight:
182.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

155-157 °C (lit.)

functional group

amine
thiourea

SMILES string

COc1ccccc1NC(N)=S

InChI

1S/C8H10N2OS/c1-11-7-5-3-2-4-6(7)10-8(9)12/h2-5H,1H3,(H3,9,10,12)

InChI key

HXCHZMHFZXNFIX-UHFFFAOYSA-N

General description

1-(2-Methoxyphenyl)-2-thiourea is a sulphur compound.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service