Skip to Content
MilliporeSigma
All Photos(2)

Documents

360902

Sigma-Aldrich

Ethyl trimethylsilyl malonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3CCH2OCOCH2COOSi(CH3)3
CAS Number:
Molecular Weight:
204.30
Beilstein:
3082383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.416 (lit.)

bp

75 °C/0.5 mmHg (lit.)

density

1 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC(=O)CC(=O)O[Si](C)(C)C

InChI

1S/C8H16O4Si/c1-5-11-7(9)6-8(10)12-13(2,3)4/h5-6H2,1-4H3

InChI key

PVXMKQLVICGFSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl trimethylsilyl malonate is an ester.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service