Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360252

Sigma-Aldrich

7-Methylxanthine

99%

Synonym(s):

2,6-Dihydroxy-7-methylpurine, Heteroxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N4O2
CAS Number:
Molecular Weight:
166.14
Beilstein:
171027
EC Number:
MDL number:

Assay

99%

SMILES string

Cn1cnc2NC(=O)NC(=O)c12

InChI

1S/C6H6N4O2/c1-10-2-7-4-3(10)5(11)9-6(12)8-4/h2H,1H3,(H2,8,9,11,12)

InChI key

PFWLFWPASULGAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service