Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

360147

Sigma-Aldrich

[1,2-Bis(diphenylphosphino)ethane]dichlorocobalt(II)

Synonym(s):

Dichloro[ethylenebis[diphenylphosphine]]cobalt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PCH2CH2P(C6H5)2]CoCl2
CAS Number:
Molecular Weight:
528.26
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: cobalt
reagent type: catalyst

mp

260 °C (dec.) (lit.)

SMILES string

Cl[Co]Cl.C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H24P2.2ClH.Co/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26;;;/h1-20H,21-22H2;2*1H;/q;;;+2/p-2

InChI key

SYTWXWRJCLAZFP-UHFFFAOYSA-L

Application

Catalyst for:
  • Regio- and stereoselective intermolecular enyne coupling
  • Cross-coupling reactions of alkyl halides with aryl Grignard reagents
  • Hydrovinylation of styrene
  • Tandem radical cyclization and cross-coupling
  • Dimethylation and hydrodechlorination
  • Addition reactions

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service