Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359947

Sigma-Aldrich

Borohydride, polymer-supported

Synonym(s):

(Polystyrylmethyl)trimethylammonium borohydride

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

form

beads

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: carbonyl reactive

storage temp.

2-8°C

SMILES string

[H][B-]([H])([H])[H].C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C[N+](C)(C)Cc3ccccc3

InChI

1S/C4H12N.BH4/c1-5(2,3)4;/h1-4H3;1H4/q+1;-1

InChI key

LSSMEJCYZMCNIN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reducing agent with greater selectivity than the corresponding alkali borohydrides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic Communications, 19, 805-805 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service