Skip to Content
MilliporeSigma
All Photos(1)

Documents

359793

Sigma-Aldrich

Z-Lys-OH

98%

Synonym(s):

Nα-(Carbobenzyloxy)-L-lysine, Nα-Cbz-L-lysine, Nα-Z-L-Lysine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4CH(NHCOOCH2C6H5)COOH
CAS Number:
Molecular Weight:
280.32
Beilstein:
2153826
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

optical activity

[α]20/D −13±2°, c = 2 in 0.2 M HCl

reaction suitability

reaction type: solution phase peptide synthesis

mp

226-231 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NCCCC[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C14H20N2O4/c15-9-5-4-8-12(13(17)18)16-14(19)20-10-11-6-2-1-3-7-11/h1-3,6-7,12H,4-5,8-10,15H2,(H,16,19)(H,17,18)/t12-/m0/s1

InChI key

OJTJKAUNOLVMDX-LBPRGKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Educt for the convenient preparation of L-α-aminoadipic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-Woo Lim et al.
Advanced healthcare materials, 8(2), e1800953-e1800953 (2018-12-15)
Powerful adjuvants to augment vaccine efficacy with a less immunogenic vaccine system are in great demand. In this study, a novel squalene-based cationic poly(amino acid) adjuvant (CASq) that elicits both cellular (Th1) and humoral (Th2) immune responses is developed. CASq
Akifumi Kato et al.
Biochemical and biophysical research communications, 546, 35-39 (2021-02-10)
Most of the currently approved therapeutic antibodies are of the immunoglobulin gamma (IgG) κ isotype, leaving a vast opportunity for the use of IgGλ in medical treatments. The incorporation of designer amino acids into antibodies enables efficient and precise manufacturing
Kimiyasu Isobe et al.
Journal of bioscience and bioengineering, 104(3), 218-223 (2007-10-30)
An oxidase catalyzing conversion of N(alpha)-benzyloxycarbonyl-L-lysine (N(alpha)-Z-L-lysine) to N(alpha)-benzyloxycarbonyl-L-aminoadipate-delta-semialdehyde (N(alpha)-Z-L-AASA) was purified from Rhodococcus sp. AIU Z-35-1, and its properties were revealed. This enzyme catalyzed an oxidative deamination of the epsilon-amino group of N(alpha)-acyl-L-lysine and the alpha-amino group of N(epsilon)-acyl-L-lysine.
Kimiyasu Isobe et al.
Chemistry & biodiversity, 7(6), 1549-1554 (2010-06-22)
The enzymes responsible for the conversion of N(alpha)-[(benzyloxy)carbonyl]-D-lysine (N(alpha)-Z-D-lysine) to N(alpha)-Z-D-aminoadipic acid (N(alpha)-Z-D-AAA) by Rhodococcus sp. AIU Z-35-1 were identified. N(alpha)-Z-D-Lysine was first converted to N(alpha)-Z-D-aminoadipic delta-semialdehyde (N(alpha)-Z-D-AASA) by D-specific amino acid deaminase, whereas N(alpha)-Z-L-lysine was converted to N(alpha)-Z-L-AASA by
A S Bajkowski et al.
The Journal of biological chemistry, 258(3), 1650-1655 (1983-02-10)
Cathepsin B has been shown to catalyze the transfer of the N alpha-benzyloxycarbonyl-L-lysyl residue from the corresponding p-nitrophenyl ester substrate to water and dipeptide nucleophiles. These reactions occurred through the formation of an acyl-enzyme intermediate. The pH dependency of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service