Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359319

Sigma-Aldrich

4-Hexylbenzoic acid

liquid crystal, 99%

Synonym(s):

p -n -Hexylbenzoic acid, p -Hexylbenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5C6H4CO2H
CAS Number:
Molecular Weight:
206.28
Beilstein:
2046454
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99%

form

liquid crystal

SMILES string

CCCCCCc1ccc(cc1)C(O)=O

InChI

1S/C13H18O2/c1-2-3-4-5-6-11-7-9-12(10-8-11)13(14)15/h7-10H,2-6H2,1H3,(H,14,15)

InChI key

CPEPWESLFZVUEP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service