Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

359289

Sigma-Aldrich

Sodium dibenzyldithiocarbamate

97%

Synonym(s):

Dibenzyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2NCSSNa
CAS Number:
Molecular Weight:
295.40
Beilstein:
4620553
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

mp

235 °C (dec.) (lit.)

SMILES string

[Na]SC(=S)N(Cc1ccccc1)Cc2ccccc2

InChI

1S/C15H15NS2.Na/c17-15(18)16(11-13-7-3-1-4-8-13)12-14-9-5-2-6-10-14;/h1-10H,11-12H2,(H,17,18);/q;+1/p-1

InChI key

MYCPTMLDBVIIES-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Shofstahl et al.
Journal of chromatographic science, 28(5), 225-229 (1990-05-01)
The United States Environmental Protection Agency (EPA) currently specifies atomic absorption spectrophotometry as the method of analysis for the priority pollutant metals. While the method is very sensitive, it does suffer from limitations. As an alternative, the use of dibenzyldithiocarbamate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service