Skip to Content
MilliporeSigma
All Photos(1)

Documents

359130

Sigma-Aldrich

2-Methyl-1,3-dithiane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10S2
CAS Number:
Molecular Weight:
134.26
Beilstein:
102766
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.560 (lit.)

bp

56-59 °C/3 mmHg (lit.)

density

1.121 g/mL at 25 °C (lit.)

SMILES string

CC1SCCCS1

InChI

1S/C5H10S2/c1-5-6-3-2-4-7-5/h5H,2-4H2,1H3

InChI key

KXROTPXCYDXGSC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1,3-dithiane is a versatile acyl anion equivalent. Role of hyperconjugation in the conformational analysis of 2-methyl-1,3-dithiane has been reported.

Application

2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal structure of potassium trifluoro (2-methyl-1, 3-dithiano) borate, K (C5H9BF3S2).
Caracelli I, et al.
Zeitschrift fur Kristallographie - New Crystal Structures, 222(3), 345-347 (2007)
Adrien Schlachter et al.
Inorganic chemistry, 57(21), 13564-13576 (2018-10-23)
A structurally unique and strongly luminescent nonporous 3D coordination polymer (CP) [Cu8I8(methyldithiane)4] n, CP3, has been prepared in a quasi-anticipated manner from 2-methyl-1,3-dithiane, L1, and CuI. This CP incorporates an unprecedented Cu8I8 cluster built upon two side-fused open cubanes. The
Douglas S Ribeiro et al.
The Journal of organic chemistry, 68(17), 6780-6787 (2003-08-16)
Full geometry optimizations were carried out at the HF/6-31G** and B3LYP/6-31G** levels for methylcyclohexane, 2-, 3-, and 4-methyltetrahydropyran, 2-, 3-, and 4-methylpiperidine, 2-, 3-, and 4-methylthiane, 2-, 4-, and 5-methyl-1,3-dioxane, and 2-, 4-, and 5-methyl-1,3-dithiane and also for S-methyl thianium.
Amos B Smith et al.
Journal of the American Chemical Society, 128(1), 66-67 (2006-01-05)
An effective one-flask multicomponent linchpin coupling protocol employing the concept of anion relay chemistry (ARC) was developed. This concept calls for addition of an anion (5) to an epoxide (6), bearing on a distal carbon a trialkyl silyl group and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service