Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

358991

Sigma-Aldrich

N-tert-Butylmethylamine

97%

Synonym(s):

N-Methyl-tert-butylamine

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$76.90
5 ML
$171.00

$76.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$76.90
5 ML
$171.00

About This Item

Linear Formula:
(CH3)3CNHCH3
CAS Number:
Molecular Weight:
87.16
Beilstein:
1730941
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.90


Please contact Customer Service for Availability

Request a Bulk Order

Assay

97%

form

liquid

refractive index

n20/D 1.395 (lit.)

bp

67-69 °C (lit.)

density

0.727 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNC(C)(C)C

InChI

1S/C5H13N/c1-5(2,3)6-4/h6H,1-4H3

InChI key

ZQGJEUVBUVKZKS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-tert-Butylmethylamine may be used in the preparation of 3-benzyl-N-tert-butyl-N,3-dimethyl-2,3-dihydrobenzofuran-6-carboxamide.[1] N-tert-Butylmethylamine (N-Methyl-tert-butylamine) may be used in the preparation of 2-(α-N-methyl-tert-butylaminobenzyl)-1-indenone.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mobile keto allyl systems. XVI. Thermal decomposition of 2-(. alpha.-N-methyl-tert-butylaminobenzyl)-1-indenone. Deamination-rearrangement.
Murray RJ and Cromwell NH.
The Journal of Organic Chemistry, 39(26), 3939-3941 (1974)
Philippe Diaz et al.
ChemMedChem, 4(10), 1615-1629 (2009-07-29)
We recently discovered and reported a series of N-alkyl-isatin acylhydrazone derivatives that are potent cannabinoid receptor 2 (CB(2)) agonists. In an effort to improve the druglike properties of these compounds and to better understand and improve the treatment of neuropathic
Karolina Klesiewicz et al.
The Journal of antibiotics, 69(11), 825-834 (2016-03-31)
A series of 20 xanthone derivatives was synthesized and evaluated for anti-Helicobacter pylori (H. pylori) activity. Qualitative and quantitative in vitro tests using the Kirby-Bauer method (agar disc-diffusion method) were performed. The tested compounds were screened against clarithromycin- and/or metronidazole-resistant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service