Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

358789

Sigma-Aldrich

Trimethyl 2-phosphonoacrylate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2P(O)C(=CH2)CO2CH3
CAS Number:
Molecular Weight:
194.12
Beilstein:
1778576
MDL number:
UNSPSC Code:
12162002

Assay

95%

contains

0.5% 2,6-di-tert-butyl-p-cresol as stabilizer

refractive index

n20/D 1.454 (lit.)

bp

90-92 °C/0.1 mmHg (lit.)

density

1.249 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(=C)P(=O)(OC)OC

Looking for similar products? Visit Product Comparison Guide

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service