Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

357731

Sigma-Aldrich

Methyl 4-chloro-2,2-dimethyl-4-pentenoate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H13ClO2
CAS Number:
Molecular Weight:
176.64
MDL number:

Assay

98%

SMILES string

COC(=O)C(C)(C)CC(Cl)=C

InChI

1S/C8H13ClO2/c1-6(9)5-8(2,3)7(10)11-4/h1,5H2,2-4H3

InChI key

NMIYZQVNXZOOPF-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service