Skip to Content
MilliporeSigma
All Photos(1)

Documents

357669

Sigma-Aldrich

7-Chloroquinaldine

99%

Synonym(s):

7-Chloro-2-methylquinoline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8ClN
CAS Number:
Molecular Weight:
177.63
Beilstein:
115318
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

74-78 °C (lit.)

SMILES string

Cc1ccc2ccc(Cl)cc2n1

InChI

1S/C10H8ClN/c1-7-2-3-8-4-5-9(11)6-10(8)12-7/h2-6H,1H3

InChI key

WQZQFYRSYLXBGP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-assisted Preparation of 3-[2-(7-chloro-2-quinolinyl) ethenyl] benzaldehyde.
ZHANG Z, et al.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 2, 012-012 (2010)
Synthesis of unsymmetrical dithioacetals: an efficient synthesis of a novel LTD4 antagonist, L-660,711.
McNamara JM, et al.
The Journal of Organic Chemistry, 54(15), 3718-3721 (1989)
Technological Progress in Synthesis of 7-chloroquinaldine as Intermediate of Anti-asthma Drug.
Dong S, et al.
Chemical Industry Times / Hua Gong Shi Kan, 6, 12-12 (2011)
Synthesis of 7-Chloroquinaldine by Skraup Reaction.
LI S, et al.
Journal of Huaihai Institute of Technology (Natural Sciences Edition) / Huai Hai Gong Xue Yuan Xue Bao (Zi Ran Ke Xue Ban ), 3, 012-012 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service