Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

35246

Sigma-Aldrich

2,6,α,α-Tetrachlorotoluene

technical, ≥95% (GC)

Synonym(s):

1,3-Dichloro-2-dichloromethylbenzene, 2,6-Dichlorobenzal chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4Cl4
CAS Number:
Molecular Weight:
229.92
Beilstein:
908809
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.588

bp

124-126 °C/16 mmHg (lit.)

density

1.520 g/mL at 20 °C (lit.)

SMILES string

ClC(Cl)c1c(Cl)cccc1Cl

InChI

1S/C7H4Cl4/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,7H

InChI key

QQPXXHAEIGVZKQ-UHFFFAOYSA-N

General description

2,6,α,α-Tetrachlorotoluene is a halogenated hydrocarbon.

Application

2,6,α,α-Tetrachlorotoluene may be used in chemical synthesis.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service