Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

349585

Sigma-Aldrich

Benzoylferrocene

≥98%

Synonym(s):

Ferrecenophenone, Ferrocenyl phenyl ketone, Ferrocenyl(phenyl)methanone, NSC 54800, (Benzoylcyclopentadienyl)cyclopentadienyliron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14FeO
CAS Number:
Molecular Weight:
290.14
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

reaction suitability

core: iron
reagent type: catalyst

mp

105-107 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.O=C([C]2[CH][CH][CH][CH]2)c3ccccc3

InChI

1S/C12H9O.C5H5.Fe/c13-12(11-8-4-5-9-11)10-6-2-1-3-7-10;1-2-4-5-3-1;/h1-9H;1-5H;

InChI key

JJHHJZCYDPLHIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Deprotonative metalation of ferrocenes
  • Oxidative chemistry
  • Reduction of ferrocenyl ketones
  • Polymerization of C-ferrocenyl-substituted phosphaalkene

Reactant for synthesis of:
  • Planat chiral triferrocenylmethane derivative
  • Chiral ferrocenyl alcohols via asymmetric transfer hydrogenation

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kevin J T Noonan et al.
Chemical communications (Cambridge, England), 35(35), 3658-3660 (2007-08-31)
The addition polymerization of a ferrocenyl-substituted P[double bond]C bond leads to new redox-active polymers with functional ferrocene and phosphine moieties.
Yong Leng Kelvin Tan et al.
Dalton transactions (Cambridge, England : 2003), 48(48), 10871-10881 (2009-12-22)
A series of compounds is described in which one of the ethyl groups in diethylstilbestrol has been replaced by a ferrocenyl substituent. Only those derivatives incorporating phenol moieties underwent isomerisation from the Z to the E form, and some of
Gandrath Dayaker et al.
Chemical communications (Cambridge, England), 46(16), 2862-2864 (2010-04-07)
A mixed lithium-cadmium amide and a combination of lithium and zinc amides were reacted with a range of ferrocenes; deprotonative mono- or dimetallation in general occurred chemoselectively at room temperature, as evidenced by subsequent quenching with iodine.
J. R. Garabatos-Perera and H. Butenschoen,
Journal of Organometallic Chemistry, 694, 2047-2052 (2009)
Y. Wu, et al.,
Tetrahedron Asymmetry, 20, 584-587 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service