Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

348902

Sigma-Aldrich

(S)-(+)-Dihydro-5-(hydroxymethyl)-2(3H)-furanone

95%

Synonym(s):

(S)-γ-Hydroxymethyl-γ-butyrolactone, (S)-4,5-Dihydro-5-hydroxymethyl-2(3H)-furanone, (S)-5-Hydroxymethyl-2-oxotetrahydrofuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8O3
CAS Number:
Molecular Weight:
116.12
Beilstein:
1680284
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

optical activity

[α]20/D +56°, c = 3 in chloroform

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.471 (lit.)

bp

110-115 °C/0.2 mmHg (lit.)

density

1.237 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC[C@@H]1CCC(=O)O1

InChI

1S/C5H8O3/c6-3-4-1-2-5(7)8-4/h4,6H,1-3H2/t4-/m0/s1

InChI key

NSISJFFVIMQBRN-BYPYZUCNSA-N

General description

Dihydro-5-(hydroxymethyl)-2(3H)-furanone is a chiral building block widely used in the synthesis of many natural products and few biologically significant compounds like anti-HIV dideoxynucleosides. It is also used in the synthesis of optically active ligands.

Application

(S)-(+)-Dihydro-5-(hydroxymethyl)-2(3H)-furanone can be used as a chiral synthon to synthesize:
  • Potent nucleoside, acetylcholine, and (+)-muricatacin analogs.
  • Mevinic acids, which are LDL cholesterol-lowering agents.
  • Adenine based anti-HIV agent named β-F-ddA.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of dihydromevinolin and a series of related 3-hydroxy-3-methylglutaryl coenzyme A reductase inhibitors
Blackwell CM, et al.
The Journal of Organic Chemistry, 57(21), 5596-5606 (1992)
Dihydro-5-(hydroxymethyl)-2(3H)-furanone
Encyclopedia of Reagents for Organic Synthesis, Second Edition (2001)
Blackwell, C.M. et al.
The Journal of Organic Chemistry, 57, 5597-5597 (1992)
A concise synthesis of anti-viral agent F-ddA, starting from (S)-dihydro-5-(hydroxymethyl)-2 (3H)-furanone
Choudhury A, et al.
Tetrahedron Letters, 44(2), 247-250 (2003)
Enantio-and diastereoisomers of 2, 4-dimethoxy-5-(2, 3-dideoxy-5-O-tritylribofuranosyl) pyrimidine. 2′, 3′-Dideoxy pyrimidine C-nucleosides by palladium-mediated glycal-aglycon coupling
Zhang HC and Daves Jr GD
The Journal of Organic Chemistry, 58(9), 2557-2560 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service