Skip to Content
MilliporeSigma
All Photos(1)

Documents

348139

Sigma-Aldrich

1-Chloropinacolone

95%

Synonym(s):

1-Chloro-3,3-dimethyl-2-butanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCH2Cl
CAS Number:
Molecular Weight:
134.60
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

liquid

refractive index

n20/D 1.442 (lit.)

bp

170-173 °C (lit.)

mp

−13 °C (lit.)

density

1.025 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)C(=O)CCl

InChI

1S/C6H11ClO/c1-6(2,3)5(8)4-7/h4H2,1-3H3

InChI key

ULSAJQMHTGKPIY-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Gas chromatographic determination of monochloropinacoline in the air, water and biological materials].
Kh T Asilbekova et al.
Gigiena i sanitariia, (1)(1), 61-62 (1992-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service