Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

347973

Sigma-Aldrich

Pentafluorophenyl chlorothionoformate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC(S)OC6F5
CAS Number:
Molecular Weight:
262.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.481 (lit.)

bp

98-102 °C/50 mmHg (lit.)

density

1.635 g/mL at 25 °C (lit.)

functional group

chloro
fluoro

SMILES string

Fc1c(F)c(F)c(OC(Cl)=S)c(F)c1F

InChI

1S/C7ClF5OS/c8-7(15)14-6-4(12)2(10)1(9)3(11)5(6)13

InChI key

DKFQHZNKNWNZCO-UHFFFAOYSA-N

Application

Pentafluorophenyl chlorothionoformate may be used:
  • as derivatizing agent during radical-chain deoxygenations of primary alcohols[1]
  • as a reagent during the conversion of ribonucleoside to arabinonucleosides[2]
  • as reagent during the conversion of 6,8-diethyl-7-hydroxy-5-propyl-hexahydro-indolizin-3-one to O-6,8-diethyl-octahydro-3-oxo-5-propylindolizin-7-yl benzothioate[3]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramaiah Kumareswaran et al.
The Journal of organic chemistry, 69(26), 9151-9158 (2004-12-22)
Starting from succinamide and 1,2-heptadiene-4-ol, a racemic allene-aldehyde substrate, 20, suitable for R(3)SiSnR'(3)-mediated cyclization was synthesized in six steps and in 21% yield. Stereoselective cyclization (relative cis configuration at the new stereogenic centers of the homoallyl alcohol generated) proceeded smoothly
The invention of radical reactions. Part XXI. Simple methods for the radical deoxygenation of primary alcohols.
Barton DHR, et al.
Tetrahedron, 47(43), 8969-8994 (1991)
Wang et al.
Organic letters, 2(2), 227-230 (2000-05-18)
[structures: see text] A regio- and stereoselective glycosylation of ribose tetraester with persilylated alloxazine to give either beta-N1 or beta-N3 nucleosides is described. The N3 product is potentially of interest as a fluorescent nucleoside and is predicted to have the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service