Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

347752

Sigma-Aldrich

Benzo-12-crown-4

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O4
CAS Number:
Molecular Weight:
224.25
Beilstein:
1246979
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:

Assay

99%

mp

42-45 °C (lit.)

SMILES string

C1COCCOc2ccccc2OCCO1

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service