347019
(−)-(R)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyl methyl ether
98%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Assay
98%
optical activity
[α]20/D −337°, c = 1 in chloroform
mp
127-128 °C (lit.)
SMILES string
[Fe].[CH]1[CH][CH][CH][CH]1.CO[C@H](C)[C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4
Application
Ligand for the synthesis of optically active binaphthyls, and ternaphthalenes via asymmetric Grignard cross-coupling.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of the American Chemical Society, 110, 8153-8153 (1988)
Tetrahedron Letters, 30, 215-215 (1989)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service