Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

346403

Sigma-Aldrich

3-(1H-Tetrazol-5-yl)-9H-thioxanthen-9-one 10,10-dioxide monohydrate

95%

Synonym(s):

Doxantrazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N4O3S · H2O
CAS Number:
Molecular Weight:
330.32
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

255 °C (dec.) (lit.)

SMILES string

[H]O[H].O=C1c2ccccc2S(=O)(=O)c3cc(ccc13)-c4nnn[nH]4

InChI

1S/C14H8N4O3S.H2O/c19-13-9-3-1-2-4-11(9)22(20,21)12-7-8(5-6-10(12)13)14-15-17-18-16-14;/h1-7H,(H,15,16,17,18);1H2

InChI key

YRXFSKFCFQRWHG-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John P Russell et al.
The Journal of pharmacology and experimental therapeutics, 368(2), 299-307 (2018-11-11)
Abdominal pain represents a significant complaint in patients with irritable bowel syndrome (IBS). While the etiology of IBS is incompletely understood, prior exposure to gastrointestinal inflammation or psychologic stress is frequently associated with the development of symptoms. Inflammation or stress-induced
B Xue et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 27(4), 550-558 (2015-03-31)
Endotoxin exposure may be followed by visceral hypersensitvity but potential mechanisms are not fully explored. We aimed to test the hypothesis that mast cells and the cyclooxygenase pathway (COX) mediate modulation of afferent nerve sensitivity following systemic endotoxin. C57Bl6 mice

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service