Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

346357

Sigma-Aldrich

4′-(Trifluoromethyl)-2-biphenylcarboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4C6H4CO2H
CAS Number:
Molecular Weight:
266.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

169-171 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)c1ccccc1-c2ccc(cc2)C(F)(F)F

InChI

1S/C14H9F3O2/c15-14(16,17)10-7-5-9(6-8-10)11-3-1-2-4-12(11)13(18)19/h1-8H,(H,18,19)

InChI key

IQOMYCGTGFGDFN-UHFFFAOYSA-N

General description

4′-(Trifluoromethyl)-2-biphenylcarboxylic acid (xenalipin) has been tested as an effective hypolipidemic agent in animal species. It has been shown to cause significant reduction in serum cholesterol and triglyceride levels in animal models and would be beneficial in therapy for hyperlipidemia. Synthesis of 4′-(trifluoromethyl)-2-biphenylcarboxylic acid (xenalipin) has been reported. Xenalipin has been synthesized in the [14C]-labelled form, with specific activity 21.0mCi/mmol, which is suitable for the metabolism and distribution studies in animals.

Application

4′-(Trifluoromethyl)-2-biphenylcarboxylic acid may be used in chemical synthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Lewis et al.
Atherosclerosis, 64(1), 27-35 (1987-03-01)
Xenalipin (4'-trifluoromethyl-2-biphenylcarboxylic acid) is a chemically novel compound which has been found to be an effective hypolipidemic agent in two animal species. Significant reductions in serum cholesterol and triglycerides were observed in cholesterol-cholic acid-fed rats following oral doses of 10-30
Synthesis of carbon-14 labelled xenalipin-a potential hypolipidemic agent.
Hill JA and Eaddy JF.
Journal of Labelled Compounds & Radiopharmaceuticals, 31(12), 1011-1017 (1992)
The Nickel (I) Catalyzed Coupling of a Diarylzinc with an Aryl Chloride in the Synthesis of Xenalipin.
Eaddy JF.
Organic preparations and procedures international, 27(3), 367-372 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service