Skip to Content
MilliporeSigma
All Photos(1)

Documents

346241

Sigma-Aldrich

Isopropyl formimidate hydrochloride

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC(=NH)OCH(CH3)2·HCl
CAS Number:
Molecular Weight:
123.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical grade

mp

108 °C (dec.) (lit.)

SMILES string

Cl.CC(C)OC=N

InChI

1S/C4H9NO.ClH/c1-4(2)6-3-5;/h3-5H,1-2H3;1H

InChI key

NWCBOWOTHFLANG-UHFFFAOYSA-N

Application

Isopropyl formimidate hydrochloride may be used in the synthesis of 5: 6-dimethyl-1-α-D-ribofuranosylbenziminazole.

Other Notes

Contains isopropyl alcohol and ammonium chloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

609. Chemistry of the vitamin B12 group. Part II. Synthesis of 5: 6-dimethyl-1-a-D-ribofuranosylbenziminazole.
Johnson AW, et al.
Journal of the Chemical Society, 3061-3066 (1953)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service