Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345709

Sigma-Aldrich

Methyl 5-oxooctadecanoate

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$428.00

$428.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$428.00

About This Item

Linear Formula:
CH3(CH2)12CO(CH2)3CO2CH3
CAS Number:
Molecular Weight:
312.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$428.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

mp

58-60 °C (lit.)

functional group

ester
ketone

SMILES string

CCCCCCCCCCCCCC(=O)CCCC(=O)OC

InChI

1S/C19H36O3/c1-3-4-5-6-7-8-9-10-11-12-13-15-18(20)16-14-17-19(21)22-2/h3-17H2,1-2H3

InChI key

BVRPGVKJUHZXTH-UHFFFAOYSA-N

General description

Methyl 5-oxooctadecanoate is an ester.

Application

Methyl 5-oxooctadecanoate may be used in the synthesis of seven-, eight- and nine-membered monocyclic, bicyclic and tricyclic ring systems.[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sequenced reactions with samarium (II) iodide. Sequential intermolecular carbonyl addition/intramolecular nucleophilic acyl substitution for the preparation of seven-, eight-, and nine-membered carbocycles.
Molander GA and Alonso-Alija C.
The Journal of Organic Chemistry, 63(13), 4366-4373 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service