Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

345687

Sigma-Aldrich

Boron trichloride solution

10 wt. % in 2-chloroethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

concentration

10 wt. % in 2-chloroethanol

density

1.258 g/mL at 25 °C

SMILES string

ClB(Cl)Cl

InChI

1S/BCl3/c2-1(3)4

InChI key

FAQYAMRNWDIXMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A Friedel-Crafts catalyst in a ready-to-use, easy-to-handle form.
For preparing methyl esters of fatty acids and for transesterification of triglycerides.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Xie et al.
Carbohydrate research, 340(3), 481-487 (2005-02-01)
Boron trichloride has been found to promote selective deprotection of 1,2- or 1,3-cis oriented secondary benzyl ethers of per-benzylated C-glycosyl derivatives. The reactivity towards BCl(3) follows the order: C-4>or=C-2>C-6>C-3 for C-glucopyranosyl derivatives and C-3>or=C-4>C-6>C-2 for C-galactopyranosyl derivatives. Preparatively useful selective
Jayaraman Selvakumar et al.
Organic & biomolecular chemistry, 8(18), 4056-4058 (2010-07-29)
Isoindoloisoquinalinone, pyrroloisoquinolinone and benzo[a]quinolizinone units are constructed via intramolecular cyclization of the methoxy substituted N-phenethylimides using BBr(3).
George W Kabalka et al.
The Journal of organic chemistry, 73(7), 2668-2673 (2008-03-06)
A boron trihalide mediated alkyne-aldehyde coupling reaction leading to stereodefined 1,3,5-triaryl-1,5-dihalo-1,4-pentadienes is described. The study led to the discovery of a direct substitution of hydroxyl groups by stereodefined alkenyl moieties using alkenylboron dihalides. During the investigation, it was also discovered
Lei Zhang et al.
Journal of combinatorial chemistry, 7(4), 622-626 (2005-07-12)
A novel and efficient microwave-assisted, BCl3-mediated coupling reaction to synthesize salicylamide structures from phenols and isocyanates is described.
Warwick J Belcher et al.
Dalton transactions (Cambridge, England : 2003), (12)(12), 1602-1614 (2008-03-13)
The reactions of boron halides with free base porphyrins under conditions where partial hydrolysis of the boron halides can occur give diboron porphyrin complexes containing BOB moieties in which each boron is bonded to two porphyrin nitrogen atoms. BF(3).OEt(2) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service