Skip to Content
MilliporeSigma
All Photos(1)

Documents

345318

Sigma-Aldrich

Polytetrahydrofuran, linear-chain polymer with oxirane

average Mn ~1,250

Synonym(s):

α-Hydro-ω-hydroxypoly(oxy-1,4-butanediyl, oxy-1,2-ethanediyl)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002

mol wt

average Mn ~1,250


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service