Skip to Content
MilliporeSigma
All Photos(1)

Documents

345261

Sigma-Aldrich

Poly(tetrahydrofuran)

average Mn ~250

Synonym(s):

α-Hydro-ω-hydroxypoly(oxy-1,4-butanediyl), Poly(1,4-butanediol), polyTHF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H(OCH2CH2CH2CH2)nOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)
<1 mmHg ( 20 °C)

form

viscous liquid

mol wt

average Mn ~250

refractive index

n20/D 1.46

mp

−14 °C

density

1 g/mL at 25 °C

SMILES string

OCCCCO

InChI

1S/C8H18O2/c1-3-5-6-10-8(4-2)7-9/h8-9H,3-7H2,1-2H3/t8-/m0/s1

InChI key

BJZYYSAMLOBSDY-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

392.0 °F - Cleveland open cup

Flash Point(C)

200 °C - Cleveland open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Nakaoka et al.
Journal of biomedical materials research, 49(1), 99-105 (1999-11-24)
Interaction of macrophages (Mos) with polyetherurethane (PEU) was investigated to clarify the role of the Mos in the early stage of tumorigenesis of PEUs. As for the inflammatory cytokines produced from Mos, the amount of tumor necrosis factor (TNF) produced
P Banu et al.
Journal of colloid and interface science, 277(2), 304-308 (2004-09-03)
Aqueous dispersions of poly(ester-imide)s [P(E-I)s] have been prepared by dispersing the P(E-I)s in water without any external solubilizing agents. P(E-I)s were prepared from anhydride-terminated polyester prepolymer and diisocyanate. The -COOH groups in the polymer were then neutralized using triethylamine and
M Renier et al.
Journal of biomaterials science. Polymer edition, 5(3), 231-244 (1993-01-01)
Poly(etherurethane urea) (PEUU) elastomers when employed as biomedical devices may be susceptible to extraction upon implantation. Four PEUU elastomers containing a single PEUU formulation, but varying in terms of their additives, were subjected to an in vitro extraction procedure. The
Jae Hyung Park et al.
Biomaterials, 23(8), 1797-1808 (2002-04-13)
In vitro protein adsorption, platelet adhesion and activation on new hydrogel surfaces, composed of poly(ethylene oxide) (PEO) and poly(tetramethylene oxide) (PTMO) or poly(dimethyl siloxane) (PDMS), were investigated. By varying PEO length (MW = 2000 or 3400), hydrophobic components (PTMO or
Jae Hyung Park et al.
Journal of biomaterials science. Polymer edition, 13(5), 527-542 (2002-08-17)
A multiblock copolymer composed of poly(ethylene oxide) (PEO) and poly(tetramethylene oxide) (PTMO) and which forms a physical hydrogel was blended with Pellethane, a commercial segmented polyurethane (SPU) developed for various biomedical devices, to provide a PEO-rich surface with improved stability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service