Skip to Content
MilliporeSigma
All Photos(1)

Documents

345040

Sigma-Aldrich

4-Bromo-2-fluorotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Br)F
CAS Number:
Molecular Weight:
189.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.529 (lit.)

solubility

water: insoluble

density

1.492 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(Br)cc1F

InChI

1S/C7H6BrF/c1-5-2-3-6(8)4-7(5)9/h2-4H,1H3

InChI key

YZFVUQSAJMLFOZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-2-fluorotoluene may be used as starting reagent in the synthesis of 4-borono-2-fluorophenylalanine, useful reagent for positron emission tomographic (PET) studies. It may be used in the synthesis of methyl 4-bromo-2-methoxybenzoate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Methyl 4-Bromo-2-methoxybenzoate.
Zhao HY and Chen BH.
Chinese Journal of Pharmaceuticals / Chung-kuo i yao kung yeh tsa chih, 39(2), 92-92 (2008)
Synthesis of 4-borono-2-fluorophenylalanine.
Kabalka GW, et al.
Organic Prep. and Proc. Int., 32(3), 290-293 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service