Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344885

Sigma-Aldrich

Piperidine thiocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2S
CAS Number:
Molecular Weight:
144.24
MDL number:

Assay

97%

SMILES string

SC#N.C1CCNCC1

InChI

1S/C5H11N.CHNS/c1-2-4-6-5-3-1;2-1-3/h6H,1-5H2;3H

InChI key

HHUXWFCTZRENLW-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service