All Photos(1)
About This Item
Linear Formula:
Cu2(OH)PO4
CAS Number:
Molecular Weight:
239.07
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Recommended Products
Assay
97%
form
powder
application(s)
battery manufacturing
SMILES string
[Cu++].O[Cu+].[O-]P([O-])([O-])=O
InChI
1S/2Cu.H3O4P.H2O/c;;1-5(2,3)4;/h;;(H3,1,2,3,4);1H2/q2*+2;;/p-4
InChI key
WCMHCPWEQCWRSR-UHFFFAOYSA-J
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nádia M Duran et al.
Journal of agricultural and food chemistry, 65(36), 7874-7884 (2017-08-18)
Nanoparticles properties such as solubility, tunable surface charges, and singular reactivity might be explored to improve the performance of fertilizers. Nevertheless, these unique properties may also bring risks to the environment since the fate of nanoparticles is poorly understood. This
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service