Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

344168

Sigma-Aldrich

Phenyl 9-acridinecarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H13NO2
CAS Number:
Molecular Weight:
299.32
MDL number:
UNSPSC Code:
12352200

Assay

98%

mp

189-191 °C (lit.)

SMILES string

O=C(Oc1ccccc1)c2c3ccccc3nc4ccccc24


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service