Skip to Content
MilliporeSigma
All Photos(1)

Documents

344109

Sigma-Aldrich

(R)-(−)-N,N-Dimethyl-1-[(S)-2-(diphenylphosphino)ferrocenyl]ethylamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28FeNP
CAS Number:
Molecular Weight:
441.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

95%

optical activity

[α]20/D −355°, c = 0.6 in ethanol

reaction suitability

reaction type: Asymmetric synthesis
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

141-143 °C (lit.)

functional group

phosphine

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2P(c3ccccc3)c4ccccc4)N(C)C

InChI

1S/C21H23NP.C5H5.Fe/c1-17(22(2)3)20-15-10-16-21(20)23(18-11-6-4-7-12-18)19-13-8-5-9-14-19;1-2-4-5-3-1;/h4-17H,1-3H3;1-5H;/t17-;;/m1../s1

InChI key

RCAFPSZHKFOLEE-ZEECNFPPSA-N

Application

Ligand used in the enantioselective vinylation of aldehydes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iio, H. et al.
Journal of the Chemical Society. Chemical Communications, 1390-1390 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service