Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

343234

Sigma-Aldrich

Niobium(IV) carbide

5 μm, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbC
CAS Number:
Molecular Weight:
104.92
EC Number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

particle size

5 μm

density

7.6 g/mL at 25 °C (lit.)

SMILES string

C#[Nb]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yingying Zhang et al.
Nanoscale, 4(7), 2268-2271 (2012-03-01)
Highly aligned carbon nanotube (CNT) ribbons were sandwiched in epitaxial superconducting NbC films by a chemical solution deposition method. The incorporation of aligned long CNTs into NbC film enhances the normal-state conductivity and improves the superconducting properties of the assembly.
Marlies De Boeck et al.
Mutagenesis, 18(2), 177-186 (2003-03-07)
Occupational exposure to hard metal dust, consisting of tungsten carbide (WC) and metallic cobalt particles (Co), is associated with an increased risk of lung cancer, while no increased risk was observed in workers exposed to Co alone. In vitro, in
S Tsuruta et al.
Shika zairyo, kikai = Journal of the Japanese Society for Dental Materials and Devices, 9(4), 617-622 (1990-07-01)
Experimental gypsum-bonded investments containing 0.5-5.0 wt% NbC were prepared by mechanical mixing of each powder. Setting and thermal expansion measurement, compressive strength and casting accuracy for Ni-Cr alloy for metal-ceramic restoration were investigated. Analysis of NbC during heating was carried
David E Grove et al.
Langmuir : the ACS journal of surfaces and colloids, 26(21), 16517-16521 (2010-09-03)
Niobium carbide nanoparticles were synthesized by flowing methane, ethylene, or acetylene gas through a plasma generated from an arc discharge between two niobium electrodes. Varying methane, ethylene, and acetylene concentrations were employed in the studies to investigate their effects on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service