Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

342319

Sigma-Aldrich

3-Chloro-6-phenylpyridazine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7ClN2
CAS Number:
Molecular Weight:
190.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

159-161 °C (lit.)

functional group

chloro
phenyl

SMILES string

Clc1ccc(nn1)-c2ccccc2

InChI

1S/C10H7ClN2/c11-10-7-6-9(12-13-10)8-4-2-1-3-5-8/h1-7H

InChI key

BUBRFWDEAVIFMV-UHFFFAOYSA-N

General description

3-Chloro-6-phenylpyridazine is an heteroaromatic compound and is investigated by cyclic voltammetry and preparative scale electrolysis in the presence and absence of carbon dioxide. Fluorination of 3-chloro-6-phenylpyridazine with KF under solvent-free conditions in the presence of a phase transfer agent, with or without microwave irradiation is reported.

Application

3-Chloro-6-phenylpyridazine may be used in the synthesis of 6-substituted phenyl-2-(3í-substituted phenylpyridazin-6í-yl)-2,3,4,5-tetrahydropyridazin-3-ones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective and efficient fluorination of chlorodiazines under solvent-free phase transfer catalysis.
Marque S, et al.
Journal of Fluorine Chemistry, 125(12), 1847-1851 (2004)
Electrochemical carboxylation of some heteroaromatic compounds.
Fuchs P, et al.
Acta Chemica Scandinavica. Series B, 35, 185-192 (1981)
Mojahidul Islam et al.
Acta poloniae pharmaceutica, 65(3), 353-362 (2008-07-24)
A series of 6-substituted phenyl-2-(3'-substituted phenyl pyridazin-6'-yl)-2,3,4,5-tetrahydropyridazin-3-ones has been synthesized. An appropriate aromatic hydrocarbon reacts with succinic anhydride in presence of AlCl3 to yield beta-aroyl propionic acid. The corresponding acid was cyclized with hydrazine hydrate to give 6-(substituted aryl)-2,3,4,5-tetrahydro-3-pyridazinone, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service