Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

342149

Sigma-Aldrich

Tetrabutylammonium hexachloroplatinate(IV)

Synonym(s):

Bis(tetrabutylammonium)hexachloroplatinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
{[CH3(CH2)3]4N}2PtCl6
CAS Number:
Molecular Weight:
892.72
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: platinum
reagent type: catalyst

mp

225 °C (dec.) (lit.)

SMILES string

Cl[Pt--](Cl)(Cl)(Cl)(Cl)Cl.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/2C16H36N.6ClH.Pt/c2*1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;;;;;;;/h2*5-16H2,1-4H3;6*1H;/q2*+1;;;;;;;+4/p-6

InChI key

SSHOYKZPYIRFFY-UHFFFAOYSA-H

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service