Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

341363

Sigma-Aldrich

(1R,2S,5R)-(+)-5-Methyl-2-(1-methyl-1-phenylethyl)cyclohexyl chloroacetate

99%

Synonym(s):

(+)-8-Phenylmenthyl chloroacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H25ClO2
CAS Number:
Molecular Weight:
308.84
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]25/D +22°, c = 1.2 in carbon tetrachloride

mp

82-84 °C (lit.)

functional group

chloro
ester
phenyl

SMILES string

C[C@@H]1CC[C@H]([C@@H](C1)OC(=O)CCl)C(C)(C)c2ccccc2

InChI

1S/C18H25ClO2/c1-13-9-10-15(16(11-13)21-17(20)12-19)18(2,3)14-7-5-4-6-8-14/h4-8,13,15-16H,9-12H2,1-3H3/t13-,15-,16-/m1/s1

InChI key

GYWWQECFQIGECM-FVQBIDKESA-N

Application

(1R,2S,5R)-(+)-5-Methyl-2-(1-methyl-1-phenylethyl)cyclohexyl chloroacetate may be used in the synthesis of (-)-8-phenyl­menthol[1][2], (+)-8-phenylmenthyl isocyanoacetate[3] and (+)-cularine.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of (+)-8-phenylmenthyl isocyanoacetate.
Solladie-Cavallo A and Quazzotti S.
Tetrahedron Asymmetry, 3(1), 39-42 (1992)
Diastereoselective synthesis of cularine alkaloids via enium ions and an easy entry to isoquinolines by aza-Wittig electrocyclic ring closure.
Rodrigues JAR, et al.
The Journal of Organic Chemistry, 69(9), 2920-2928 (2004)
A diastereoselective radical cyclisation approach to pyroglutamates.
Goodall K and Parsons AF.
Tetrahedron Letters, 38(3), 491-494 (1997)
1R, 2S, 5R)-5-Methyl-2-(1-methyl-1-phenylethyl) cyclohexyl chloroacetate.
Sieder F, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(6), o2291-o2292 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service