Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

340588

Sigma-Aldrich

Tetrahydro-3-furanmethanol

contains 250 ppm BHT as inhibitor, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O2
CAS Number:
Molecular Weight:
102.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

viscous liquid

contains

250 ppm BHT as inhibitor

refractive index

n20/D 1.456 (lit.)

bp

76-77 °C/4 mmHg (lit.)

density

1.061 g/mL at 25 °C (lit.)

functional group

ether
hydroxyl

SMILES string

OCC1CCOC1

InChI

1S/C5H10O2/c6-3-5-1-2-7-4-5/h5-6H,1-4H2

InChI key

PCPUMGYALMOCHF-UHFFFAOYSA-N

General description

Tetrahydro-3-furanmethanol was the main product formed during hydrolysis of 1,3-dioxolane compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5- hydrolysis (2-halo-ethyl) -1,3-dioxane :Studies on Hydrolysis of 5- (2-haloethyl) -1, 3- Dioxolane.
Hou R-Z, et al.
Journal of Jilin University (Science Edition) / Chi Lin Ta Hsueh Hsueh Pao (Li Hsueh Pan ), 4, 94-95 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service