Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

340359

Sigma-Aldrich

Tributyl(phenylethynyl)tin

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C≡CSn[(CH2)3CH3]3
CAS Number:
Molecular Weight:
391.18
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
form:
liquid
Assay:
95%

Assay

95%

form

liquid

refractive index

n20/D 1.532 (lit.)

density

1.116 g/mL at 25 °C (lit.)

functional group

phenyl

SMILES string

CCCC[Sn](CCCC)(CCCC)C#Cc1ccccc1

InChI

1S/C8H5.3C4H9.Sn/c1-2-8-6-4-3-5-7-8;3*1-3-4-2;/h3-7H;3*1,3-4H2,2H3;

InChI key

PYMPTRMDPJYTDF-UHFFFAOYSA-N

Application

Tributyl(phenylethynyl)tin can be used:
  • As a substrate in the palladium-catalyzed homocoupling of organostannanes to yield biaryl compounds.[1]
  • To prepare various enynals, which are employed in the synthesis of substituted benzene derivatives catalyzed by nickel.[2]
  • In one of the key synthetic steps for the preparation of β-alkynylcorroles.[3]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient Synthesis of beta-Alkynylcorroles
Stefanelli M, et al.
European Journal of Organic Chemistry, 2015(31), 6811-6816 (2015)
A multicomponent approach to substituted benzenes involving sequential nickel-catalyzed reactions
ElDouhaibi AS, et al.
Tetrahedron, 62(49), 11460-11469 (2006)
Palladium-iminophosphine-catalyzed homocoupling of alkynylstannanes and other organostannanes using allyl acetate or air as an oxidant
Shirakawa E, et al.
Journal of Organometallic Chemistry, 670(1-2), 132-136 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service