Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

340286

Sigma-Aldrich

Dimethyl methylmalonate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(CO2CH3)2
CAS Number:
Molecular Weight:
146.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor pressure

0.2 mmHg ( 20 °C)

Assay

99%

refractive index

n20/D 1.414 (lit.)

bp

176-177 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(C)C(=O)OC

InChI

1S/C6H10O4/c1-4(5(7)9-2)6(8)10-3/h4H,1-3H3

InChI key

LRBPFPZTIZSOGG-UHFFFAOYSA-N

General description

Dimethyl methylmalonate is an ester. Microwave-enhanced Pd(0)/acetic acid catalyzed allylation of C, N and O-nucleophiles with dimethyl methylmalonate has been reported.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave-enhanced Pd (0)/acetic acid catalyzed allylation reactions of C, N, and O -pronucleophiles with alkynes.
Patil NT, et al.
Tetrahedron Letters, 45(46), 8497-8499 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service