Skip to Content
MilliporeSigma
All Photos(1)

Documents

340219

Sigma-Aldrich

5,6-Dichloropyridine-3-carboxylic acid

96%

Synonym(s):

5,6-Dichloronicotinic acid, 5,6-Dichloropyridine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2NO2
CAS Number:
Molecular Weight:
192.00
Beilstein:
383740
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

164-168 °C (lit.)

SMILES string

OC(=O)c1cnc(Cl)c(Cl)c1

InChI

1S/C6H3Cl2NO2/c7-4-1-3(6(10)11)2-9-5(4)8/h1-2H,(H,10,11)

InChI key

RNRLTTNKVLFZJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5,6-Dichloropyridine-3-carboxylic acid is a dihalonicotinic acid.

Application

5,6-Dichloropyridine-3-carboxylic acid (5,6-Dichloronicotinic acid) was used in the preparation of 5-chloro-6-iodonicotinic acid by iodine displacement.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Some 2, 5-and 5, 6-dihalonicotinic acids and their precursors. IV.
Setliff FL and Lane JE.
Journal of Chemical and Engineering Data, 21(2), 246-247 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service