Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339911

Sigma-Aldrich

2-Hydroxy-3-isopropyl-6-methylbenzoic acid

98%

Synonym(s):

3-Isopropyl-6-methylsalicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2[CH(CH3)2](CH3)CO2H
CAS Number:
Molecular Weight:
194.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

127-129 °C (lit.)

functional group

carboxylic acid

SMILES string

CC(C)c1ccc(C)c(C(O)=O)c1O

InChI

1S/C11H14O3/c1-6(2)8-5-4-7(3)9(10(8)12)11(13)14/h4-6,12H,1-3H3,(H,13,14)

InChI key

FNWNGQGTFICQJU-UHFFFAOYSA-N

General description

2-Hydroxy-3-isopropyl-6-methylbenzoic acid is also referred as o-thymotic acid. Analgesic and antistress evaluation of o-thymotic acid in rat has been reported.

Application

2-Hydroxy-3-isopropyl-6-methylbenzoic acid is suitable for nuclear reactor applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of the tri-o-thymotide-ethyl methyl sulfoxide (2: 1) clathrate. Crystallographic estimation of the enantioselectivity.
Allemand J and Gerdil R.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 38(8), 2312-2315 (1982)
Analgesic and anti-inflammatory evaluation of thymotic acid and certain homologs.
F J Marozzi et al.
Journal of pharmaceutical sciences, 57(6), 989-994 (1968-06-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service