Skip to Content
MilliporeSigma
All Photos(1)

Documents

339806

Sigma-Aldrich

3,5-Dimethoxybenzylamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH2NH2
CAS Number:
Molecular Weight:
167.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

refractive index

n20/D 1.542 (lit.)

bp

94-96 °C/0.1 mmHg (lit.)

mp

35-37 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

COc1cc(CN)cc(OC)c1

InChI

1S/C9H13NO2/c1-11-8-3-7(6-10)4-9(5-8)12-2/h3-5H,6,10H2,1-2H3

InChI key

YGZJTYCCONJJGZ-UHFFFAOYSA-N

General description

3,5-Dimethoxybenzylamine was prepared by LiAlH4-reduction of 3,5-dimethoxybenzaldoxime.

Application

3,5-Dimethoxybenzylamine was used in the preparation of trisammonium tris(hexafluoro phosphate) salt. It was also used in the synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jishan Wu et al.
Proceedings of the National Academy of Sciences of the United States of America, 104(44), 17266-17271 (2007-10-20)
In this article, we report on the efficient synthesis of well defined, homogeneous [n]rotaxanes (n up to 11) by a template-directed thermodynamic clipping approach. By employing dynamic covalent chemistry in the form of reversible imine bond formation, [n]rotaxanes with dialkylammonium
Methoxy-substituted benzyl isothiocyanates and N-benzyl thioureas.
Bach ERIK and Kjaer A.
Acta Chemica Scandinavica, 25(7) (1971)
The exclusivity of multivalency in dynamic covalent processes.
Jovica D Badjić et al.
Angewandte Chemie (International ed. in English), 43(25), 3273-3278 (2004-06-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service