Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339229

Sigma-Aldrich

Samarium cobalt

powder

Synonym(s):

Cobalt compound with samarium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm2Co7
CAS Number:
Molecular Weight:
713.25
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

composition

Sa:Co, 2-7

reaction suitability

reagent type: catalyst
core: samarium

SMILES string

[Co].[Sm]

InChI

1S/Co.Sm

InChI key

KPLQYGBQNPPQGA-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elliot J Crooks et al.
The Journal of biological chemistry, 295(27), 8914-8927 (2020-05-08)
Cerebral amyloid angiopathy (CAA) is a vascular disorder that primarily involves deposition of the 40-residue-long β-amyloid peptide (Aβ40) in and along small blood vessels of the brain. CAA is often associated with Alzheimer's disease (AD), which is characterized by amyloid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service