Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339091

Sigma-Aldrich

Silver hexafluoroarsenate(V)

95%

Synonym(s):

Arsenic silver fluoride (AgAsF6 ), Silver hexafluoroarsenate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgAsF6
CAS Number:
Molecular Weight:
296.78
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Assay

95%

form

powder and chunks

SMILES string

[Ag+].F[As-](F)(F)(F)(F)F

InChI

1S/Ag.AsF6/c;2-1(3,4,5,6)7/q+1;-1

InChI key

UNSKNPXMHZPWPB-UHFFFAOYSA-N

Application

Used to prepare silver 3,3′-dicyanodiphenylacetylene coordination networks for study of the effect of ligands on network structure toward the goal of engineering novel materials.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith A. Hirsch et al.
Inorganic chemistry, 36(14), 2960-2968 (1997-07-02)
Coordination networks of 3,3'-dicyanodiphenylacetylene (3,3'-DCPA, 1) with silver(I) salts characterized by single-crystal X-ray analysis are described. Network topology is found to depend on both the counterion and solvent employed during crystallization. The conformation adopted by the ligand varies between planar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service