Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

338605

Sigma-Aldrich

Methyl 3-hydroxy-5-isoxazolecarboxylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NO4
CAS Number:
Molecular Weight:
143.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

160-163 °C (lit.)

SMILES string

COC(=O)c1cc(O)no1

InChI

1S/C5H5NO4/c1-9-5(8)3-2-4(7)6-10-3/h2H,1H3,(H,6,7)

InChI key

BBFWUUBQSXVHHZ-UHFFFAOYSA-N

Application

Methyl 3-hydroxy-5-isoxazolecarboxylate was used in the enantioselective synthesis of a key precursor to the tetracycline antibiotics. It was also used in the preparation of formamidinopiperidine analog, an N-amidinopiperidine compound.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C B Xue et al.
Bioorganic & medicinal chemistry letters, 8(24), 3499-3504 (1999-02-06)
Despite the excellent in vitro potency of a series of benzamide glycoprotein IIb/IIIa antagonists, which have been reported previously, poor in vivo potency in the inhibition of platelet aggregation was observed when the most potent inhibitor XU057 was dosed intravenously
Jason D Brubaker et al.
Organic letters, 9(18), 3523-3525 (2007-08-19)
A practical, enantioselective synthetic route to a key precursor to the tetracycline antibiotics is reported. The route proceeds in nine steps (21% yield) from the commercial substance methyl 3-hydroxy-5-isoxazolecarboxylate. Key steps in the route involve enantioselective addition of divinylzinc to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service