Skip to Content
MilliporeSigma
All Photos(1)

Documents

338362

Sigma-Aldrich

2,3,4-Trifluoronitrobenzene

99%

Synonym(s):

1,2,3-Trifluoro-4-nitrobenzene, 4-Nitro-1,2,3-trifluorobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2NO2
CAS Number:
Molecular Weight:
177.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

92 °C/20 mmHg (lit.)

density

1.541 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(F)c(F)c1F

InChI

1S/C6H2F3NO2/c7-3-1-2-4(10(11)12)6(9)5(3)8/h1-2H

InChI key

ARCACZWMYGILNI-UHFFFAOYSA-N

General description

2,3,4-Trifluoronitrobenzene is a fluoronitrobenzene and its biotransformation under methanogenic conditions has been studied by semicontinuous and batch tests. 2,3,4-Trifluoronitrobenzene is an important pharmaceutical intermediate.

Application

2,3,4-Trifluoronitrobenzene was employed as starting reagent in the synthesis of ofloxacin. It was also used in the synthesis of the third generation quinolones antibacterial drugs.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrasonic Promoted in Synthesis of 2, 3, 4-trichloronitrobenzene [J].
Yunqiang L and Guohui C.
Guangzhou Chemical Industry/ Guangzhou Huagong, 3, 024-024 (2009)
Analytical profile of the fluoroquinolone antibacterials. I. Ofloxacin.
Okeri HA and Arhewoh IM.
African Journal of Biotechnology, 7(6) (2008)
Zhiqing Zhao et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 49(10), 1187-1197 (2014-05-23)
The fluorinated compounds are becoming a ubiquitous class of environmental contaminants because of their widespread applications, and their fate is a matter of great concern under anaerobic environment. In this work, the biotransformation of five fluoronitrobenzenes (FNBs), i.e., 2-fluoronitrobenzene (2-FNB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service