Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

338354

Sigma-Aldrich

3-Phenoxyaniline

98%

Synonym(s):

3-Aminophenyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4NH2
CAS Number:
Molecular Weight:
185.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

329-330 °C (lit.)

mp

41-44 °C (lit.)

functional group

phenoxy

SMILES string

Nc1cccc(Oc2ccccc2)c1

InChI

1S/C12H11NO/c13-10-5-4-8-12(9-10)14-11-6-2-1-3-7-11/h1-9H,13H2

InChI key

UCSYVYFGMFODMY-UHFFFAOYSA-N

Application

3-Phenoxyaniline was used in the preparation of 4-phenoxyaniline-3′-bromopropionamide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dona Chianelli et al.
Bioorganic & medicinal chemistry, 11(23), 5059-5068 (2003-11-08)
Nonpeptide compounds that mimic bioactive peptides are desirable for a number of clinical indications. We report a new practical method for the design of scaffolds exhibiting drug-like properties that are suitable for the display of peptide pharmacophores. The synthesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service