Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

338028

Sigma-Aldrich

1,4-Cyclohexanedimethanol diglycidyl ether, mixture of cis and trans

technical grade

Synonym(s):

1,4-Bis(glycidoxymethyl)cyclohexane, 1,4-Bis(hydroxymethyl)cyclohexane diglycidyl ether, 1,4-Bis[(2,3-epoxypropoxy)methyl]cyclohexane, 1,4-Bis[(glycidyloxy)methyl]cyclohexane, 1,4-Cyclohexanedimethanol diglycidyl ether, 2,2′-[1,4-Cyclohexanediylbis(methyleneoxymethylene)]bis[oxirane]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O4
CAS Number:
Molecular Weight:
256.34
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Pricing and availability is not currently available.

grade

technical grade

vapor density

>1 (vs air)

vapor pressure

11.4 mmHg ( 25 °C)

refractive index

n20/D 1.481 (lit.)

density

1.1 g/mL at 25 °C (lit.)

SMILES string

C1CC(CCC1COCC2CO2)COCC3CO3

InChI

1S/C14H24O4/c1-2-12(6-16-8-14-10-18-14)4-3-11(1)5-15-7-13-9-17-13/h11-14H,1-10H2

InChI key

VQMQXWYQIIUJIT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service